Human Recombinant IL-17F (from E. coli)
Supplier: VWR International
VWR100-152-100UG VWR100-152-1MG VWR100-152-25UG VWR100-152-500UG VWR100-152-5UG
77750-496EA 600.95 USD
77750-496 77750-498 77750-500 77750-502 77750-504
Human Recombinant IL-17F (from E. coli)
Proteins and Peptides
Interleukin 17F (IL-17F), a member of the IL-17 cytokine family, is secreted by activated CD4+ T cells and monocytes.  IL-17F binds the IL-17 receptor related molecule, IL17RC, to promote the production of the interleukin 6 (IL-6), interleukin 8 (IL-8), and granulocyte macrophage colony-stimulating factor (GM-CSF) cytokines.  IL-17F also functions to regulate matrix turnover rates, inhibit endothelial cell angiogenesis, and induce the endothelial cell expression of interleukin 2 (IL-2), monocyte chemoattractant protein-1 (MCP-1), and transforming growth factor beta 1 (TGF-b1).  
  • High quality
  • Low endotoxin
  • Affordable
Precaución:
For research use only.