4-Amino-1-((3aR,4R,6R,6aR)-6-(hydroxymethyl)-2,2-dimethyltetrahydrofuro[3,4-d][1,3]dioxol-4-yl)pyrimidin-2(1H)-one ≥98%

Supplier: Ambeed
Warning

A793438-1G
77749-566EA 36 USD
77749-566
4-Amino-1-((3aR,4R,6R,6aR)-6-(hydroxymethyl)-2,2-dimethyltetrahydrofuro[3,4-d][1,3]dioxol-4-yl)pyrimidin-2(1H)-one ≥98%
4-Amino-1-((3aR,4R,6R,6aR)-6-(hydroxymethyl)-2,2-dimethyltetrahydrofuro[3,4-d][1,3]dioxol-4-yl)pyrimidin-2(1H)-one

Precaución: for Research Use Only

Formula: C₁₂H₁₇N₃O₅
MW: 283.28 g/mol
MDL Number: MFCD15145298
CAS Number: 362-42-5
New Product

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR