6-Bromo-2,4-dichloropyrrolo[2,1-f][1,2,4]triazine ≥98%

Supplier: Ambeed
Warning

A776196-250MG
77749-532EA 154 USD
77749-532
6-Bromo-2,4-dichloropyrrolo[2,1-f][1,2,4]triazine ≥98%
6-Bromo-2,4-dichloropyrrolo[2,1-f][1,2,4]triazine

Precaución: for Research Use Only

Formula: C₆H₂BrCl₂N₃
MW: 266.91 g/mol
MDL Number: MFCD12400791
CAS Number: 1160995-23-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR