2-Pentyn-1-ol ≥98%

Supplier: Ambeed
Danger

A701929-5G
77749-440EA 26 USD
77749-440
2-Pentyn-1-ol ≥98%
2-Pentyn-1-ol

Precaución: for Research Use Only

Formula: C₅H₈O
MW: 84.12 g/mol
MDL Number: MFCD00040915
CAS Number: 6261-22-9
UN: 1987
ADR: 3,Ⅲ
New Product

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR