4-Pentylpyridine ≥98%

Supplier: Ambeed
Warning

Synonyms: 4-Amylpyridine, 4-Pentylpyridine

A648101-5G
77749-334EA 328 USD
77749-334
4-Pentylpyridine ≥98%
4-Pentylpyridine

Precaución: for Research Use Only

Formula: C₁₀H₁₅N
MW: 149.24 g/mol
MDL Number: MFCD02093422
CAS Number: 2961-50-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR