1-(Oxetan-3-yl)ethan-1-one ≥97%

Supplier: Ambeed
Warning

A179614-100MG
77748-173EA 204 USD
77748-173
1-(Oxetan-3-yl)ethan-1-one ≥97%
1-(Oxetan-3-yl)ethan-1-one

Precaución: for Research Use Only

Formula: C₅H₈O₂
MW: 100.12 g/mol
MDL Number: MFCD30072292
CAS Number: 1507872-90-3
New Product

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR