3-Bromo-1,8-naphthyridine ≥98%

Supplier: Ambeed
Warning

A145827-100MG
77748-019EA 117 USD
77748-019
3-Bromo-1,8-naphthyridine ≥98%
3-Bromo-1,8-naphthyridine

Precaución: for Research Use Only

Formula: C₈H₅BrN₂
MW: 209.04 g/mol
MDL Number: MFCD04113794
CAS Number: 17965-78-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR