4-Bromo-3-fluoro-2-methoxyaniline ≥97%

Supplier: Ambeed
Warning

Synonyms: 4-Bromo-3-fluoro-o-anisidine, 6-Amino-3-bromo-2-fluoroanisole

A757662-1G
77747-492EA 268 USD
77747-492
4-Bromo-3-fluoro-2-methoxyaniline ≥97%
4-Bromo-3-fluoro-2-methoxyaniline

Precaución: for Research Use Only

Formula: C₇H₇BrFNO
MW: 220.039 g/mol
MDL Number: MFCD22570403
CAS Number: 1137869-95-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR