2-Bromo-3-fluoropyrazine ≥95%

Supplier: Ambeed
Warning

A162877-1G
77746-108EA 583 USD
77746-108
2-Bromo-3-fluoropyrazine ≥95%
2-Bromo-3-fluoropyrazine

Precaución: for Research Use Only

Formula: C₄H₂BrFN₂
MW: 176.97 g/mol
MDL Number: MFCD15144418
CAS Number: 206278-27-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR