SKI-II 98%

Supplier: Ambeed
Warning

Synonyms: 4-((4-(4-Chlorophenyl)thiazol-2-yl)amino)phenol, SKI II

A122860-10MG A122860-1MG A122860-25MG A122860-5MG A122860-1G A122860-100MG A122860-250MG A122860-50MG
77246-176EA 43 USD
77246-176 77246-178 77246-180 77246-182 77571-394 77745-897 77745-898 77745-899
SKI-II 98%
SKI-II

Precaución: for Research Use Only

Formula: C₁₅H₁₁ClN₂OS
MW: 302.776 g/mol
MDL Number: MFCD00733553
CAS Number: 312636-16-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR