Doxylamine Hydrogen Succinate, LoGiCal, LGC Standards

Supplier: LGC Standards

LGCFOR0101.00
77697-059EA 206.65 USD
77697-059
Doxylamine Hydrogen Succinate, LoGiCal, LGC Standards
Standards Organic Standards
New Product

Order Now

SPECIFICATIONS

CAS No. 562-10-7
Formula C₂₁H₂₈N₂O₅
Molecular Weight 388.464 g/mol

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR