Doxapram Hydrochloride Monohydrate 1.0 mg/ml in Methanol (as anhydrous free base), LoGiCal, LGC Standards

Supplier: LGC Standards

LGCAMP0525.00-01
77696-907EA 109.65 USD
77696-907
Doxapram Hydrochloride Monohydrate 1.0 mg/ml in Methanol (as anhydrous free base), LoGiCal, LGC Standards
Standards Organic Standards

Order Now

SPECIFICATIONS

CAS No. 7081-53-0
Formula C₂₄H₃₀N₂O₂·HCl·H₂O
Molecular Weight 432.98 g/mol

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR