Vanadium Standard: V @ 10 µg/mL in 2% HNO3, VHG, LGC Standards
Supplier: LGC Standards
VHG-LVN-100
77695-350EA 60 USD
77695-350
Vanadium Standard: V @ 10 µg/mL in 2% HNO3, VHG, LGC Standards
Standards Cationic Standards Vanadium Standards