Multi-Element 22 Components Calibration Standard for 6020 CLP-M: 10 µg/mL Ag,Al,As,Ba,Be,Ca,Cd,Co,Cr,Cu,Fe,K,Mg,Mn,Na,Ni,Pb,Sb,Se,Tl,V,Zn in 2% HNO3,tr. Tartaric Acid,VHG,LGC Standards
Supplier: LGC Standards
VHG-LCAL6020-100
77693-154EA 296.05 USD
77693-154
Multi-Element 22 Components Calibration Standard for 6020 CLP-M: 10 µg/mL Ag,Al,As,Ba,Be,Ca,Cd,Co,Cr,Cu,Fe,K,Mg,Mn,Na,Ni,Pb,Sb,Se,Tl,V,Zn in 2% HNO3,tr. Tartaric Acid,VHG,LGC Standards
Standards Cationic Standards Multi-Element Standards