(3R)-3-(Bromomethyl)pyrrolidine, N-BOC protected ≥96%, Ambeed.Inc

Supplier: Ambeed
Warning

Synonyms: tert-Butyl (3R)-3-(bromomethyl)pyrrolidine-1-carboxylate, (R)-1-Boc-3-(Bromomethyl)pyrrolidine

A945400-250MG A945400-100MG A945400-5G
77614-660EA 92.7 USD
77614-660 77684-891 77684-892
(3R)-3-(Bromomethyl)pyrrolidine, N-BOC protected ≥96%, Ambeed.Inc
(3R)-3-(Bromomethyl)pyrrolidine, N-BOC protected

Precaución: For research use only.

Formula: C₁₀H₁₈BrNO₂
MW: 264.159 g/mol
MDL Number: MFCD08059335
CAS Number: 1067230-65-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR