(5,6-Difluoropyridin-3-yl)boronic acid ≥96%

Supplier: Ambeed
Warning

A705831-1G
77684-619EA 317 USD
77684-619
(5,6-Difluoropyridin-3-yl)boronic acid ≥96%
(5,6-Difluoropyridin-3-yl)boronic acid

Precaución: For research use only.

Formula: C₅H₄BF₂NO₂
MW: 158.89 g/mol
MDL Number: MFCD21609554
CAS Number: 1366482-40-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR