BioReady Gold Colloid Passive Conjugation Kit
Supplier: NANOCOMPOSIX, INC.
PCKR
77680-436EA 760 USD
77680-436
BioReady Gold Colloid Passive Conjugation Kit
Assays Immunological Assays
Use the BioReady passive conjugation kit to conjugate any proteins to 40 nm and 80 nm bare citrate OD 20 gold particles.
Our conjugation kit provides the flexibility to conjugate at any OD from 1 to 20.
Información de suministro: Kit compenents: BioReady 40 nm bare citrate gold, OD 20, 5 ml, bioready 80 nm bare citrate gold, OD 20, 5 ml, titration buffers covering pH 6 - 9.7, 1 ml each, blocking buffer, 5 ml, conjugate diluent, 5 ml, 10% NaCl for salt test, 5 ml