Ultra Uniform Gold Nanospheres with a PEG-Carboxyl surface, nanoComposix
Supplier: NANOCOMPOSIX, INC.
AUXU50-10M AUXU50-1M AUXU50-30M AUXU50-5M AUXU10-30M AUXU10-5M AUXU20-10M AUXU20-1M AUXU20-30M AUXU20-5M AUXU30-10M AUXU30-1M AUXU30-30M AUXU30-5M AUXS100-25M AUXS30-25M AUXS50-25M AUXU100-10M AUXU100-1M AUXU100-30M AUXU100-5M AUXU10-10M AUXU10-1M
77680-202EA 684 USD
77680-202 77680-204 77680-206 77680-208 77680-182 77680-184 77680-186 77680-188 77680-190 77680-192 77680-194 77680-196 77680-198 77680-200 77680-164 77680-166 77680-168 77680-170 77680-172 77680-174 77680-176 77680-178 77680-180
Ultra Uniform Gold Nanospheres with a PEG-Carboxyl surface, nanoComposix
Beads Microcarriers
Ultra Uniform Gold Nanospheres for applications requiring the highest uniformity in size and shape
  • For applications requiring the highest uniformity in size and shape
Our Ultra Uniform particles have the lowest coefficients of variation available anywhere. They are used as particle size and concentration standards, electron microscopy standards, and for applications where tight tolerances on the particle size, shape, mass, or volume are required. Each batch is extensively characterized to meet highest quality standards, ensuring reliable andMore Product Information