Flexible Polyurethane Feeding Tubes for Rodents
Supplier: INSTECH LABORATORIES, INC
FTPU-13-88 FTPU-13-88-50 FTPU-16-38 FTPU-16-38-50
77679-308BX 2067.55 USD
77679-308 77679-310 77679-312 77679-314
Flexible Polyurethane Feeding Tubes for Rodents
Animal Supplies Animal Feeding Needles
Disposable polyurethane animal feeding needles for mice and rats. Used for viscous compounds.
  • Larger inner diameter
  • Straight rounded tip
  • Clear medical-grade copolyester hub
  • Sterilized by ETO gas
  • Manufactured in USA
These tubes feature a larger inner diameter for easier delivery of viscous compounds and a straight rounded tip that wipes clean to help in cases where aspiration of compounds could be hazardous to the animal. The tube is made of soft polyurethane and the hub is clear medical-grade copolyester.
Available in two tube sizes: 16 G (5 Fr)More Product Information