Educational Microscope with Eyepiece Pointers and Koehler Illumination, DM750, Leica Microsystems
Supplier: Leica Microsystems
8122676
77676-420EA 1996.7 USD
77676-420
Educational Microscope with Eyepiece Pointers and Koehler Illumination, DM750, Leica Microsystems
Microscopes
The Leica DM750 microscope offers educational excellence with its versatile design. It includes an interactive eyepiece pointer, Koehler illumination, and precision optics for clear observations. Elevate your classroom experience with this powerful and user-friendly microscope.
  • User-friendly design and innovative features provide a reliable and technologically advanced microscope to inspire and engage students in the exploration of the microscopic world
  • Perfect light: LED illumination designed to provide even lighting across the full field of view without
More Product Information