1-(Methylamino)propan-2-ol ≥95%

Supplier: Ambeed
Danger

A663482-5G
77673-814EA 376 USD
77673-814
1-(Methylamino)propan-2-ol ≥95%
1-(Methylamino)propan-2-ol

Precaución: for Research Use Only

Formula: C₄H₁₁NO
MDL Number: MFCD00089199
CAS Number: 16667-45-1
UN: 2735
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR