Methyl-2,3-dihydro-1,4-benzodioxine-5-carboxylate ≥95%

Supplier: Ambeed
Warning

Synonyms: Methyl 2,3-dihydrobenzo[b][1,4]dioxine-5-carboxylate

A475341-5G
77673-228EA 906 USD
77673-228
Methyl-2,3-dihydro-1,4-benzodioxine-5-carboxylate ≥95%
Methyl-2,3-dihydro-1,4-benzodioxine-5-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₁₀O₄
MW: 194.19 g/mol
MDL Number: MFCD01546256
CAS Number: 214894-91-4
New Product

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR