2-Chloro-4-hydrazinopyridine ≥95%

Supplier: Ambeed
Warning

A453839-1G
77673-136EA 28 USD
77673-136
2-Chloro-4-hydrazinopyridine ≥95%
2-Chloro-4-hydrazinopyridine

Precaución: for Research Use Only

Formula: C₅H₆ClN₃
MW: 143.58 g/mol
MDL Number: MFCD08460247
CAS Number: 700811-29-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR