Methyl 4-amino-6-methylnicotinate ≥95%

Supplier: Ambeed
Warning

A161371-100MG
77671-280EA 116 USD
77671-280
Methyl 4-amino-6-methylnicotinate ≥95%
Methyl 4-amino-6-methylnicotinate

Precaución: for Research Use Only

Formula: C₈H₁₀N₂O₂
MW: 166.18 g/mol
MDL Number: MFCD07374917
CAS Number: 886372-01-6
New Product

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR