Reusable Rapid Punch Biopsy Kits
Supplier: WORLD PRECISION INSTRUMENTS LLC
504639 WP1212 WP3030 WP2020F WP1010
77663-718EA 264.86 USD
77663-718 77665-596 77665-700 77665-616 77665-574
Reusable Rapid Punch Biopsy Kits
Biopsy Punches
The Rapid-Punch consists of a razor sharp cutting tip designed to cut, retrieve and store cored samples from source materials such as tissue, gels, paper, cloth, leaves, paint chips, films or other thin substrates.
  • Heavy duty reusable punch
  • Designed for heavy, numerous and precise sampling operations up to 2000 punches
  • Full CNC machined 6061-T6 Aluminum alloy punch parts
  • Tip composition: Stainless steel 440C with heat treatment HRC 58 hardness
  • All new intelligent plunger system and changeable plunger sizes
  • Sterilization: ETO (ethyline oxide)
  • Kit includes:
More Product Information