Linear Stimulus Isolators, A395
Supplier: WORLD PRECISION INSTRUMENTS LLC
A395RC SYS-A395D SYS-A395R
77664-000EA 5284.09 USD
77664-000 77665-182 77665-184
Linear Stimulus Isolators, A395
Electrical Meters Voltmeters
A395 generates a user-defined output current of wave shape; DC, AC, pulse and combinations. Battery operated, photoelectrically isolated from the input voltage drive, the instrument regenerates output currents which are linearly proportional to the analog voltage waveforms provided by your D/A converter or signal generator.
  • Creates a constant current replica of analog waveforms
  • Amplitude of the output current is voltage controlled
  • Input voltage from –10 to +10 V
  • Three current ranges from 100 μA to 10 mA
  • Built-in test resistors
  • Digital display shows current
More Product Information
New Product
Order Now
SPECIFICATIONS
Environmentally Preferable