High Current Stimulus Isolators, A385
Supplier: WORLD PRECISION INSTRUMENTS LLC
A385RC SYS-A385R
77663-998EA 5220.74 USD
77663-998 77665-180
High Current Stimulus Isolators, A385
Electrical Meters Voltmeters
The A385 is an optically isolated current source, which can generate up to 100 mA of unipolar or biphasic constant current pulses or DC. Pulse duration is controlled manually or by an external 5 V command. Output current amplitude is determined by a 3-digit 10-turn potentiometer. Maximum output voltage between the stimulating electrodes is +36 V.
  • Constant current to 100 mA
  • Unipolar or bipolar stimulation modes
  • Built-in non-compliance alarm
  • Input is optically isolated
  • Standard TTL triggering
  • DC test mode
  • Powered by six rechargeable lead acid batteries
  • 36 V
More Product Information