Liquid Waveguide Capillary Cells
Supplier: WORLD PRECISION INSTRUMENTS LLC
LWCC-3050 LWCC-3100 LWCC-3250 LWCC-3500
77664-546EA 4773.52 USD
77664-546 77664-548 77664-550 77664-552
Liquid Waveguide Capillary Cells
Flow Cells
UV/VIS/NIR absorbance spectroscopy is governed by Beer’s Law, where the absorbance signal is proportional to chemical concentration, light path length and the compound’s specific molar absorption coefficient.
  • Optical sample flow cell that combines an increased optical pathlength with a small sample volume
  • Measure liquids in a continuous flow or using discrete samples
  • Connects with 600 µm core optical fibers to fiber optic spectrometers and light sources via SMA terminations
  • Efficient measurement of low-volume or low-concentration (ppb-ppt) aqueous samples
More Product Information