Hydrogen Sulfide Sensors, World Precision Instruments
Supplier: WORLD PRECISION INSTRUMENTS LLC
ISO-H2S-100-CXX ISO-H2S-2
77664-464PK 3180.67 USD
77664-464 77664-466
Hydrogen Sulfide Sensors, World Precision Instruments
Electrodes Dissolved Oxygen Electrodes
Low detection limit sensors to record in vitro. Hydrogen sulfide is generally thought of in terms of a poisonous gas, it is endogenously produced in many mammalian tissues. It has been detected in micromolar amounts in blood and brain tissue.
  • A dry sensor that works like a 2 mm wet sensor
  • Response time: ~5 s
  • Detection limit: <5 nM
  • Sensitivity: 1 to 4 pA/nM
  • Hight selectivity and sensitivity to hydrogen sulfide with fast response time
  • Broad linearity
Hydrogen sulfide is reported as having a broad range of biological functions and although its
More Product Information
New Product
Order Now
SPECIFICATIONS
Environmentally Preferable
Response Time ~5 <5 sec