Modular Amplifer Blank Channel Panel
Supplier: WORLD PRECISION INSTRUMENTS LLC
74050
77663-950EA 242.87 USD
77663-950
Modular Amplifer Blank Channel Panel
Power Supplies
The ISDB chassis holds up to eight modules and includes a power supply. You may include any combination of BRIDGE8 Transducer Amplifier modules or ISO-DAM8A single channel modules.
If you choose fewer than eight modules, you need to order the ISDB blank panels to fill the empty slots in the chassis. Please specify line voltage when ordering.
When configuring your ISDB system:
Order a chassis and power supply enclosure
Select up to 8 modules
Order blank panels to fill empty slots
Información para pedidos: The modules are sold separately.