Modular Amplifer Chassis
Supplier: WORLD PRECISION INSTRUMENTS LLC
74030
77663-946EA 5543.44 USD
77663-946
Modular Amplifer Chassis
Power Supplies
Photo illustrates a complete amplifier system and is shown with the chassis and eight ISO-DAM8A amplifier modules.
  • Order a chassis and power supply enclosure
  • Select up to 8 modules
  • Order blank panels to fill empty slots
  • Chassis accepts combination of bioamplifiers and transducer amplifiers
  • Flexible channel count (1 to 8) allows expandability
  • Notch filter targets AC line noise sources
  • Variable gain output amplitude
  • Wide ±10 V output range