Trough Filament for P30 Micropipette
Supplier: WORLD PRECISION INSTRUMENTS LLC
505119
77663-834EA 179.51 USD
77663-834
Trough Filament for P30 Micropipette
Puller and Inserter Tools
A glass puller is a laboratory instrument used to create fine, tapered glass micropipettes or microelectrodes. The choice of filament for the glass puller is crucial, as it directly influences the heating element's performance and, consequently, the quality of the pulled glass. Different types and shapes of filaments can be used, and the selection depends on factors such as the desired pipette. A trough filament is used in some capillary glass pullers, and it is designed to creates a trough or indentation along its length.
This design allows for localized heating. The groove is somewhat U-More Product Information