Digital Caliper
Supplier: WORLD PRECISION INSTRUMENTS LLC
501601
77662-644EA 207.89 USD
77662-644
Digital Caliper
Calipers
The high quality electronic digital caliper is a useful tool - no laboratory should be without one because it is more accurate and easier to use than the traditional analog devices. Measure in either inches or millimeters at the touch of a button. The floating zero feature allows you to read the increment without calculation.
  • Measures up to 150 mm
  • 0.01 mm resolution
The caliper is made from hardened stainless. The caliper is designed to be water-resistant to IP54 as defined in the IEC529 standard. The IP54 code’s first digit '5' means dust-protected.More Product Information