Cuticle Nipper
Supplier: WORLD PRECISION INSTRUMENTS LLC
500137
77662-134EA 63.15 USD
77662-134
Cuticle Nipper
Cutters
Ring handled cuticle nippers are a handy instrument to have in a life science laboratory. While they are designed for trimming excess cuticles, hangnails, and dead skin around the nails, the unique shape makes them versatile.
  • Ring handles for greater control
  • Angled blades for greater visibility and precision cuts
When working with plants or animals, you can precisely trim sections of tissue in tight spots. You can clip the ends of sutures, cleanly prune young plants, trim away exoskeleton component during microdissection, or collect tissue samples.More Product Information