5-Iodo-2-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

19590-5G 19590-25G 19590-1G 19590-500MG
77634-282EA 442 USD
77634-282 77654-064 77641-114 77649-164
5-Iodo-2-methylbenzaldehyde ≥97%
5-Iodo-2-methylbenzaldehyde

Precaución: For Research Use Only

Formula: 246.045 g/mol
MW: C₈H₇IO
Storage Temperature: Ambient
MDL Number: MFCD18397777
CAS Number: 65874-26-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR