2-(4-Chlorophenyl)thiophene ≥97%

Supplier: AOBChem USA

Warning

80227-5G 80227-1G 80227-500MG
77630-680EA 1680 USD
77630-680 77640-008 77653-086
2-(4-Chlorophenyl)thiophene ≥97%
2-(4-Chlorophenyl)thiophene

Precaución: For Research Use Only

Formula: 194.681 g/mol
MW: C₁₀H₇ClS
Storage Temperature: Ambient
MDL Number: MFCD06802538
CAS Number: 40133-23-1
New Product

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR