3-((tert-Butyldiphenylsilyl)oxy)-2,2-dimethylpropan-1-ol ≥95%

Supplier: Ambeed
Warning

A1705251-1G
77625-742EA 31.93 USD
77625-742
3-((tert-Butyldiphenylsilyl)oxy)-2,2-dimethylpropan-1-ol ≥95%
3-((tert-Butyldiphenylsilyl)oxy)-2,2-dimethylpropan-1-ol

Precaución: for Research Use Only

Formula: C₂₁H₃₀O₂Si
MW: 342.55 g/mol
MDL Number: MFCD34187028
CAS Number: 147509-22-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR