Guardian™ 5000 Hotplates and Stirrers
Supplier: Ohaus
30541629 30981776 30981778 30981780 30981782 30541631 30541633 30541635 30541637
77623-708EA 871.25 USD
77623-708 77623-732 77623-734 77623-736 77623-738 77623-710 77623-712 77623-714 77623-716
Guardian™ 5000 Hotplates and Stirrers
Stirrers Magnetic Hotplate Stirrers
The next generation Guardian 5000TM Hotplate Stirrers are engineered for safety and versatility across research, academic and industrial settings. They offer powerful heating and mixing, with premium hardware and software to support diverse users safely and reliably. Heating models reach up to 550 °C and feature three temperature control modes for quick heating or precise heating without overshoot. Select from five models with three plate sizes.
  • Operation: 100 to 120 VAC or 220 to 240 VAC 50/60 Hz power entry, rotary switch knobs control
  • Communication: RS23
More Product Information