Dimethyl phenylboronate ≥97% (by HPLC)

Supplier: Ambeed
Warning

A1439840-100MG
77622-066EA 45.32 USD
77622-066
Dimethyl phenylboronate ≥97% (by HPLC)
Dimethyl phenylboronate

Precaución: for Research Use Only

Formula: C₈H₁₁BO₂
MW: 149.9827 g/mol
MDL Number: MFCD24857381
CAS Number: 13471-35-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR