4,7-Dibromo-5,6-difluoro-2-(heptadecan-9-yl)-2H-benzo[d][1,2,3]triazole ≥98%

Supplier: Ambeed
Warning

A1164068-1G
77620-328EA 241.02 USD
77620-328
4,7-Dibromo-5,6-difluoro-2-(heptadecan-9-yl)-2H-benzo[d][1,2,3]triazole ≥98%
4,7-Dibromo-5,6-difluoro-2-(heptadecan-9-yl)-2H-benzo[d][1,2,3]triazole

Precaución: for Research Use Only

Formula: C₂₃H₃₅Br₂F₂N₃
MW: 551.3489 g/mol
CAS Number: 1616860-69-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR