Workbrutes® G2 Over Large Work Boots
Supplier: Tingley Rubber
45850-2X 45850-LG 45850-MD 45850-SM 45850-XL 45850-XS
77619-240EA 35.69 USD
77619-240 77619-242 77619-244 77619-246 77619-248 77619-250
Workbrutes® G2 Over Large Work Boots
Shoes and Boots
Workbrutes® G2 are wider and have more room in the toe for the ideal fit over safety toe work boots and other large shoes, while high-quality PVC provides ultimate protection from liquids, mud and harsh chemical environments.
  • 17" height overshoe
  • Seamless injection-molding for 100% liquidproof protection
  • Expansion pleats for easy donning even over work boots
  • Thumb pocket at the top allows a non-slip grip for easy on
  • Frigiflex® PVC compound stays stretchy and flexible in cold tempearture
  • Increased thickness in the heel, toe and cleats for longer life
  • No
More Product Information