VWR® Dissolved Oxygen Electrodes, Avantor
Supplier: VWR International
DO1 DO3
77618-816EA 453.2 USD
77618-816 77618-818
VWR® Dissolved Oxygen Electrodes, Avantor
Electrodes Dissolved Oxygen Electrodes
The VWR® dissolved oxygen electrodes allow for the measurement of oxygen within a solution.
  • Optical or polarographic sensor technology options
  • Built-in temperature probe
  • For use with VWR® dissolved oxygen meters
For application specific recommendations, please reference "VWR® electrode selection guide" located in the documents section.