VWR® Premium Series Upright Ultra-Low Temperature Freezers, 230 V / 50 Hz
Supplier: VWR International
77618-806 77618-770 77618-782 77618-794
77618-806EA 16858.38 USD
77618-806 77618-770 77618-782 77618-794
VWR® Premium Series Upright Ultra-Low Temperature Freezers, 230 V / 50 Hz
Freezers Ultra Low Temperature Freezers
The VWR® Premium Series ULT freezers provide the power of performance with energy efficiency for a greener tomorrow.
  • 230 V / 50 Hz models
  • Operate between -50 to -86 °C
  • 4 storage capacities: (400, 500, 600, 700 2" cryoboxes)
  • ENERGY STAR® certified
  • Easy-to-use touch button interface
  • Vacuum insulation panels and high density water-blown foam
  • USB for easy export of recorded temperatures and events
  • 5 year warranty parts and labor
Experience the power of these premium ULT freezers, meticulously engineered to excel in the most demanding laboratory settings.
More Product Information
New Product
Order Now
SPECIFICATIONS
Doors 1 outer, 2 inner doors 1 outer, 2 inner doors 1 outer, 2 inner doors 1 outer, 2 inner doors
Model Premium ULT freezer 700 Premium ULT freezer 400 Premium ULT freezer 500 Premium ULT freezer 600
Environmentally Preferable
Electrical 100-230 V, 50/60 Hz, EU specific country cord 100-230 V, 50/60 Hz, EU specific country cord 100-230 V, 50/60 Hz, EU specific country cord 100-230 V, 50/60 Hz, EU specific country cord
Shelves/drawers 4 shelves (3 adjustable) 4 shelves (3 adjustable) 4 shelves (3 adjustable) 4 shelves (3 adjustable)
Door Lock Key lock with padlock hasps Key lock with padlock hasps Key lock with padlock hasps Key lock with padlock hasps
Plug Country specific cord Country specific cord Country specific cord Country specific cord