4-Bromo-6-phenyldibenzo[b,d]furan ≥97%

Supplier: Ambeed
Warning

A548079-1G
77617-340EA 20.6 USD
77617-340
4-Bromo-6-phenyldibenzo[b,d]furan ≥97%
4-Bromo-6-phenyldibenzo[b,d]furan

Precaución: for Research Use Only

Formula: C₁₈H₁₁BrO
MW: 323.1833 g/mol
MDL Number: MFCD31556237
CAS Number: 1010068-84-4
New Product

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR