[1-(2,2,2-Trifluoroethyl)-1H-pyrazol-5-yl]methanol ≥95%

Supplier: Ambeed
Warning

Synonyms: (1-(2,2,2-Trifluoroethyl)-1H-pyrazol-5-yl)methanol

A395995-250MG
77616-708EA 56.65 USD
77616-708
[1-(2,2,2-Trifluoroethyl)-1H-pyrazol-5-yl]methanol ≥95%
[1-(2,2,2-Trifluoroethyl)-1H-pyrazol-5-yl]methanol

Precaución: for Research Use Only

Formula: C₆H₇F₃N₂O
MW: 180.128 g/mol
MDL Number: MFCD15976765
CAS Number: 1296225-26-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR