3-Bromo-2,6-dihydroxybenzoic acid 97%

Supplier: Ambeed
Warning

A112253-1G A112253-5G A112253-25G A112253-10G
77189-134EA 11 USD
77189-134 77189-136 77570-678 77614-878
3-Bromo-2,6-dihydroxybenzoic acid 97%
3-Bromo-2,6-dihydroxybenzoic acid

Precaución: for Research Use Only

Formula: C₇H₅BrO₄
MW: 233.02 g/mol
MDL Number: MFCD03788827
CAS Number: 26792-49-4
New Product

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR