SafeMask® Architect Pro™ Surgical/N95 Respirator, Medicom
Supplier: Medicom
203214 203314 203414
77603-742BX 60.03 USD
77603-742 77603-744 77603-746
SafeMask® Architect Pro™ Surgical/N95 Respirator, Medicom
Respirators Disposable Respirators Particulate Respirators
Medical grade respirator providing optimal respiratory protection by filtering 95% of particulate aerosols free of oil.
  • High-quality materials and duck-bill shape
  • Lightweight, breathable construction
  • Unique single split comfort band
  • Adjustable nose piece
This medical grade respirator has been evaluated for use in healthcare settings according to recognized standards for biocompatibility, flammability, and fluid resistance. It provides optimal respiratory protection by filtering 95% of particulate aerosols free of oil. High-quality materials andMore Product Information