Methyl 2-methyloxirane-2-carboxylate ≥97%

Supplier: Ambeed
Danger

Synonyms: Methyl 2-methylglycidate

A804226-1G
77603-040EA 22.55 USD
77603-040
Methyl 2-methyloxirane-2-carboxylate ≥97%
Methyl 2-methyloxirane-2-carboxylate

Precaución: for Research Use Only

Formula: C₅H₈O₃
MW: 116.12 g/mol
MDL Number: MFCD00198125
CAS Number: 58653-97-7
UN: 3272
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR