6-Fluoro-4-chromanone ≥99%

Supplier: Ambeed
Warning

Synonyms: 6-Fluorochroman-4-one

A271835-5G
77601-164EA 15.38 USD
77601-164
6-Fluoro-4-chromanone ≥99%
6-Fluoro-4-chromanone

Precaución: for Research Use Only

Formula: C₉H₇FO₂
MW: 166.15 g/mol
MDL Number: MFCD00038654
CAS Number: 66892-34-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR