Allyltriethylsilane ≥98%

Supplier: Ambeed
Danger

A769979-5G
77599-060EA 59.45 USD
77599-060
Allyltriethylsilane ≥98%
Allyltriethylsilane

Precaución: for Research Use Only

Formula: C₉H₂₀Si
MW: 156.34 g/mol
MDL Number: MFCD18207713
CAS Number: 17898-21-4
UN: 1993
ADR: 3 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR